Learn More
MilliporeSigma™ Supelco™ SUPELCOSIL™ LC-PCN HPLC Column, 5 µm Particle Size
The LC-PCN cyanopropyl bonded phase columns are preconditioned for fast and reliable analyses of tricyclic antidepressants.
Manufacturer:  MilliporeSigma™ Supelco™ 59189
 View more versions of this product
Catalog No. 11-100-3150
Add to cart
Specifications
Specifications
Yes
Reversed Phase
20 cm
120 Å
HPLC
SUPELCOSIL
Cyano Phase/Silica Gel, Spherical Particle Platform
4.6 mm
5 μm
170 m2/g
Cyano
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC