Learn More
MilliporeSigma Supelco SupelMIP SPE Tube - NNAL 96-Well plate, Bed. Wt. 25 mg/well
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
Manufacturer: MilliporeSigma Supelco 53255U
Description
SupelMIP SPE phases were developed by MIP Technologies AB, which is one of the leading authorities and commercial pioneers of molecular imprinted polymers for process scale separations, analytical chromatography, and sample preparation. The SupelMIP SPE line consist s of highly cross-linked polymers that are engineered to extract a single analyte of interest or a class of structurally related analytes with an extremely high degree of selectivity. This is possible because selectivity is introduced during MIP synthesis in which a template molecule, designed to mimic the analyte, guides the formation of specific cavities or imprints that are sterically and chemically complementary to the analyte(s) of interest.
By careful design of the imprinting site, either by molecular modeling, experimental design, or screening methods, the binding cavities can be engineered to offer multiple interaction points with the analyte(s) of interest. This leads to a stronger interaction between the solid-phase and the analyte(s). As a consequence, harsher wash conditions can be tolerated during SPE methodology resulting in cleaner extracts. Because extraction selectivity is significantly improved, lower background is observed allowing analysts to achieve lower limits of detection.
- Highly selective SPE for trace analysis in complex matrices.
- Achieve lower detection limits.
- Significant time and cost savings.
- Improved MS-compatibility
Specifications
96-Well SPE Plate | |
25 mg/well | |
SPE | |
SupelMIP |
Polypropylene | |
NNAL | |
1/Pk. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
SupelMIP is a registered trademark of Sigma-Aldrich Co. LLC