Learn More
Thermo Scientific™ Super Clean™Gas Cartridge Filter Baseplates and Accessories
Description
Use quality baseplates/manifolds for your Thermo Scientific™ Super Clean™ Cartridge Filters. A range of baseplate options allows configuration to your individual requirements. Needle valves ensure that the gas line is not contaminated during cartridge change. The baseplates have a one-time installation procedure. Super Clean Cartridge Filters are not included.
- Use with Super Clean cartridge filters
- Filters not included
Specifications
Specifications
| For Use With (Equipment) | Triple Filter |
| Description | For Gas: Carrier Gas Only |
| Type | Single Base |