Thermo Scientific™ Super Clean™ Gas Cartridge Filters
Improve gas quality and optimize GC analysis with easy-to-use Thermo Scientific™ Super Clean™ Gas Cartridge Filters.
Manufacturer: Thermo Scientific™ 60180822
 View more versions of this product
Catalog No. 60180822
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Indicator Change
    Technique
    Capacity (Triple) 1.8g H2O, 500mL O2, 7g HCs (as n-butane)
    (Combi) 3.5g H2O, 12g HCs (as n-butane)
    Type 3 Cartridge Pack – contains 1 x Triple Filter and 2 x Combi Filter (Moisture, Hydrocarbons)
    Cartridge Type Triple, Combi (Moisture, Hydrocarbons)
    Item Description Baseplate not included
    For Use With (Triple) Inert carrier, He, H2, N2, AR
    (Combi) Inert carrier, He, H2, N2, AR, Air
    Ensure high-purity (99.9999% or 6.0 grade) output gas for optimal GC performance using Thermo Scientific™ Super Clean™ Gas Cartridge Filters. The baseplates can be configured to individual user needs, and there is no contamination during cartridge change. Easy-to-use and cost-effective, Super Clean Gas Cartridge Filters enable fast, tool-free replacement.
  • Description & Specifications
Related Products
Product Content Feedback