Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metro™ Super Erecta 4 in. High Stackable Ledge for Wire Shelving

Super Erecta 4 in. High Stackable Ledges for Wire Shelving prevent items from protruding or falling from shelves.

$30.30 - $71.70

Specifications

Autoclavable Not Autoclavable
Color Gray
Material Epoxy-coated Steel
Type Stackable Ledge for Wire Shelving
For Use With (Application) All Laboratory and Clean Room Non-Autoclavable Applications
View More Specs

Products 11
Catalog Number Mfr. No. Length (English) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Price Quantity & Availability  
13-460-321
View Documents
Metro™
L14N4K4
14 in.
Each for $30.30
Add to cart
 
13-460-318
View Documents
Metro™
L18N4K4
18 in.
Each for $31.90
Add to cart
 
13-460-319
View Documents
Metro™
L21N4K4
21 in.
Each for $32.95
Add to cart
 
13-460-320
View Documents
Metro™
L24N4K4
24 in.
Each for $36.10
Add to cart
 
13-460-325
View Documents
Metro™
L30N4K4
30 in.
Each for $40.90
Add to cart
 
13-460-317
View Documents
Metro™
L36N4K4
36 in.
Each for $45.70
Add to cart
 
13-460-324
View Documents
Metro™
L42N4K4
42 in.
Each for $55.80
Add to cart
 
13-460-316
View Documents
Metro™
L48N4K4
48 in.
Each for $55.80
Add to cart
 
13-460-326
View Documents
Metro™
L54N4K4
54 in.
Each for $59.50
Add to cart
 
13-460-323
View Documents
Metro™
L60N4K4
60 in.
Each for $59.50
Add to cart
 
13-460-322
View Documents
Metro™
L72N4K4
72 in.
Each for $71.70
Add to cart
 
Description

Description

  • Only compatible with wire shelving.
  • Ledges may be stacked to form an 8 in. (203 mm) or higher barrier.
Specifications

Specifications

Not Autoclavable
Epoxy-coated Steel
All Laboratory and Clean Room Non-Autoclavable Applications
139.7 mm
1.621 in.
Gray
Stackable Ledge for Wire Shelving
Super Erecta Shelving
4 in.
41.1734 mm
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit