Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metro™ Super Erecta Wall Mount Shelving Post with Brackets

Combine with shelves and shelf supports to create a post-type wall mounted shelving solution with Super Erecta Wall Mount Shelving Posts with Brackets.

$63.20 - $98.80

Specifications

Autoclavable Not Autoclavable
Load Bearing Capacity (English) 250 lb.
Color Gray
Material Epoxy-coated Steel
For Use With (Application) All Laboratory and Clean Room Non-Autoclavable Applications
View More Specs

Products 4
Catalog Number Mfr. No. Height (English) Price Quantity & Availability  
Catalog Number Mfr. No. Height (English) Price Quantity & Availability  
13-460-268
View Documents
Metro™
13PDFK4
13 in.
Each for $63.20
Add to cart
 
13-460-266
View Documents
Metro™
33PDFK4
33 in.
Each for $94.50
Add to cart
 
13-460-269
View Documents
Metro™
54PDFK4
54 in.
Each for $97.70
Add to cart
 
13-460-267
View Documents
Metro™
63PDFK4
63 in.
Each for $98.80
Add to cart
 
Description

Description

  • 13 in. posts are ideal for 1-shelf tier configurations.
  • 33 in. posts support 2-shelf configurations.
  • 54 in. posts can accommodate 3 or more shelving tiers.
  • 63 in. posts are designed to handle 4 or more shelving tiers.
Specifications

Specifications

Not Autoclavable
Gray
All Laboratory and Clean Room Non-Autoclavable Applications
2 in.
250 lb.
Epoxy-coated Steel
Super Erecta Shelving
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit