Learn More
Thermo Scientific™ Super-Nuova™ Multi-Position Digital Stirrers
Carry out solubility studies, digestion, titration and enzymatic studies with Thermo Scientific? Super-Nuova? Multi-Position Digital Stirrers.
Manufacturer:  Thermo Scientific™ S13603033Q
 View more versions of this product
Catalog No. 11-475-33Q
  Check Availability
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Specifications
Description
Carry out solubility studies, digestion, titration and enzymatic studies with Thermo Scientific™ Super-Nuova™ Multi-Position Digital Stirrers. Offering four individually-controlled stirring positions, ceramic tops and 4-function memory, these stirrers are designed for stronger coupling and stirring control with advanced safety features. Ideal for labs with multiple users.
Key Features
  • Thermo Scientific™ Stir Trac™ stir control provides improved slow-speed stirring, consistent speed, and strong magnetic coupling — reducing the risk of stir bar decoupling
  • Microprocessor control ensures accurate and stable setpoints for stirring speed
  • Stir Trac braking feature brings stir bar to an immediate stop for quick flask removal
  • Single control knob controls speed; lock feature prevents accidental changes
  • Digital rpm display
  • Clean easily and resist alkalis and acids
  • Seamless, reflective white ceramic surface aids sample visibility
  • Stirring adjustable in units of 1rpm
  • RS-232 port outputs time count, stir speed (set point) directly to your PC
Safety Features
  • Rugged low-profile cast-aluminum body provides stability and durability
  • Unit design protects internal components from accidental spills
  • Walk-away timer shuts off stirring after 1, 2, 4, 8, or 12 hr.
Includes:
  • Detachable line cord and plug
Description & Specifications
Specifications
50 to 1,200rpm
220/240V
10.2cm
41.1cm
28.7cm
50/60Hz
9.3kg
220/240V 50/60Hz
Stirrers
5
4 in.
16.2 in.
11.3 in.
4 Position, 220-240V
20.5 lb.
cCSAus (all models), CE (220-240V models)
16.2 x 11.3 x 4 in. (41.1 x 28.7 x 10.2cm)
Accessories