Learn More
Thermo Scientific™ Super-Nuova™ Single-Position Digital Stirrers
Obtain unmatched performance, safety and control with Thermo Scientific? Super-Nuova? Single-Position Digital Stirrers.
Manufacturer:  Thermo Scientific™ S13393033Q
 View more versions of this product
Catalog No. 09-047-120Q
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
Obtain unmatched performance, safety and control with Thermo Scientific™ Super-Nuova™ Single-Position Digital Stirrers, which deliver easy operation and sophisticated controls for maximum safety. Thermo Scientific™ StirTrac™ technology offers improved coupling and stronger magnetic control. Ceramic-top general-purpose stirrers with four-function memory are ideal for labs with multiple users.
Operating Features
  • Microprocessor control ensures accurate and stable setpoints for stirring speed
  • Single control knob controls speed; lock feature prevents accidental changes
  • Digital rpm display
  • Seamless, reflective white ceramic surface aids sample visibility
  • Stirring adjustable in units of 1rpm
  • RS-232 port outputs time count, stir speed (setpoint) directly to PC
Safety Features
  • Rugged low-profile cast-aluminum body provides stability and durability
  • Unit design protects internal components from accidental spills
  • Walk-away timer shuts off stirring after 1, 2, 4, 8, or 12 hrs.
Includes:
  • Detachable line cord and plug
Description & Specifications
Specifications
50 to 1,200rpm
6L
Ceramic
10.2cm
41.1cm
28.7cm
10.25 sq. in.
60Hz
cCSAus (all models), CE (220-240V models)
16.2 x 11.3 x 4 in. (41.1 x 28.7 x 10.2cm)
1
220/240V
4 in.
16.2 in.
11.3 in.
10.25 x 10.25, Single Position
26cm²
10.25 x 10.25, Single Position
220/240V 60Hz
Stirrers
Accessories