Learn More
ECOLAB INC SUPER TRUMP, 4/CS DFS Item
Manufacturer:  ECOLAB INC 6112740 X
Super Trump is a concentrated, high-alkaline, heavy duty liquid detergent for machine warewashing. It can also be used for Bar Glass Washing, Automatic Duct Cleaning Systems and can be automatically dispensed directly from the shipping container. The concentrated, high-alkaline formula strips away heavy grease, protein and starch food soils that ordinary detergents can miss and is effective in light, medium and heavy soil conditions. It also contains special water conditioning agents to help reduce spotting and streaking.
Catalog No. NC0140895