Thermo Scientific™ SuperChip™ Microarray Slides

Choose from several standard surface chemistries and custom coatings with Thermo Scientific™ SuperChip™ Microarray Slides.

Manufacturer: Thermo Scientific™ C62-5591-M20

 View more versions of this product

Catalog No. 12-550-58

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Color Plain
    Coating Aldehyde Silane
    Description SuperChip Microarray Slides
    Enhanced Surface Yes
    Length (Metric) 70mm
    Width (Metric) 20mm

    Choose from several standard surface chemistries and custom coatings with Thermo Scientific™ SuperChip™ Microarray Slides. Slides are manufactured from high-quality, ultra-flat, low-fluorescence glass.

    Aminopropylsilane-coated slides are ideal for microarrays fabricated with PCR products or proteins as probes;

    Poly-L-Lysine-coated slides offer an economical alternative to aminopropylsilane;

    Aldehyde-silane-coated slides provide a uniform coating of aldehydes for the attachment of DNA and protein molecules;

    Epoxysilane-coated slides utilize a coating of epoxide groups for the attachment of amine-modified DNA, proteins, peptides, etc. with an amine end. Also recently added,

    SuperChip AdvaNCe, nitrocellulose-coated slides for use in reverse phase arrays or protein microarray. For multiplexing applications, choose

    SmartPlex multiplexing microarray platform. SmartPlex allows for easy printing of arrays onto glass prior to attaching 96-well upper structure for multiplexing.

    • Consistent surface coating to provide a uniform DNA or protein immobilizing surface
    • Bind an optimal amount of probe while minimizing nonspecific binding of labeled targets
    • Many available with an enhanced surface (ES) treatment that produces a more uniform, slightly larger spot size without changing surface chemistry
    • Compatible with standard microarray spotting and detection systems
    • Ready for immediate use or may be stored at room temperature for up to a year (six months for Poly-L-Lysine)
    • Overall dimensions: 25
    • Printable area: 20 x 70mm
    • Packaged in a secure no-contact mBox™ and vacuum sealed in a foil barrier bag

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit