Learn More
Thermo Scientific™ SuperChip™ Microarray Slides Available on GSA/VA Contract for Federal Government customers only.
Choose from several standard surface chemistries and custom coatings with Thermo Scientific™ SuperChip™ Microarray Slides.
Manufacturer:  Thermo Scientific™ C505588M20
 View more versions of this product
Catalog No. 12-550-55
  Check Availability
Add to cart
Specifications
Description
Choose from several standard surface chemistries and custom coatings with Thermo Scientific™ SuperChip™ Microarray Slides. Slides are manufactured from high-quality, ultra-flat, low-fluorescence glass.
Aminopropylsilane-Coated Slides:
  • Ideal for microarrays fabricated with PCR products or proteins as probes
Poly-L-Lysine-Coated Slides:
  • Offer an economical alternative to aminopropylsilane
Aldehyde-Silane-Coated Slides:
  • Provide a uniform coating of aldehydes for the attachment of DNA and protein molecules
Epoxysilane-Coated Slides:
  • Utilize a coating of epoxide groups for the attachment of amine-modified DNA, proteins, peptides, etc. with an amine end
SuperChip Advance:
  • Nitrocellulose-coated slides for use in reverse phase arrays or protein microarray
SmartPlex:
  • Multiplexing microarray platform SmartPlex allows for easy printing of arrays onto glass prior to attaching 96-well upper structure for multiplexing
Product Details:
  • Consistent surface coating to provide a uniform DNA or protein immobilizing surface
  • Optimal amount of probe binded while minimizing nonspecific binding of labeled targets
  • Many available with an enhanced surface (ES) treatment that produces a more uniform, slightly larger spot size without changing surface chemistry
  • Compatible with standard microarray spotting and detection systems
  • Ready for immediate use or may be stored at room temperature for up to a year (six months for Poly-L-Lysine)
  • Printable area: 20 x 70mm
  • Secure no-contact mBox™ packaging, vacuum sealed in a foil barrier bag
Description & Specifications
Specifications
Epoxysilane
No
20mm
SuperChip Microarray Slides
70mm
Plain