Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Superdex™ 200 Increase 10/300 GL Prepacked Tricorn™ Columns: PROMO

For size exclusion chromatography and analysis of proteins with molecular weights between 10,000 and 600,000. GE Healthcare Lifescience™ Superdex™ 200 Increase 10/300 GL Prepacked Tricorn™ Columns are designed for small scale preparative purification.

Manufacturer:  GE Healthcare 28990944PM

Catalog No. 45-002-570LR

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • For small-scale (25 to 500μL) preparative purification and for analytical purposes, suited for antibodies
  • Allows characterization of proteins, aggregate analysis of antibodies, size homogeneity analysis of membrane proteins, or protein-protein interactions
  • Prepacked with Superdex 200 Increase media
  • Composite of cross-linked high-flow agarose and dextran base matrix for high flow rates
  • Smaller bead size of average 8.6μm for increased resolution
  • Flow Rate: <1.80mL/min1
  • Bed Volume: Approximately 24mL
  • Column 1.D.: 10mm
  • Columns are supplied with two fingertight connectors 1/16 in. male for connection to AKTA™ or other system

Specifications

Specifications

25 to 500μL
Superdex 200 Increase 10/300 GL Prepacked Column
Small-scale (25 to 500μL) preparative purification and analysis
Cross-linked agarose and dextran
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit