Learn More
GE Healthcare Superdex 75 Increase Prepacked Column, 10/300 GL: PROMO
Use these columns in preparative purification as well as for characterization and analysis of proteins with molecular weights between Mr 3,000 and 70,000, such as recombinant tagged proteins. GE Healthcare HisTrap™ excel Prepacked Columns feature tolerance to repeated harsh cleaning procedures at high pH ensures long column lifetime and minimizes carryover.
Manufacturer:  GE Healthcare 29148721PM
Catalog No. 45-003-050LR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Up to 50% higher resolution compared with predecessor Superdex 75, for improved purity and analysis results
  • Runtime reduced down to one third compared with predecessor Superdex 75, with maintained resolution for results faster
  • Versatile use in both preparative and analytical applications, especially with recombinant tagged proteins
  • Tolerance to repeated harsh cleaning procedures at high pH ensures long column lifetime and minimizes carryover
  • Higher lot-to-lot consistency compared with predecessor Superdex 75
Specifications
Specifications
High resolution size exclusion chromatography (SEC) in small scale (25 to 500μL sample volume)
4° to 30°C
≈24mL