Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Clutch Gear™ Impact-Resistant Mechanics Gloves with PVC Sure Grip™

These gloves fully protect the back of the hand from impacts, without sacrificing dexterity or flexibility. PVC Sure Grip™ palm patches enhance grip capabilities.

$17.23 - $47.75

Specifications

Color Yellow, Black
Cut Resistance Rating ANSI A2
For Use With (Application) Mechanics, Construction, Oil/Gas Drilling and Material Handling
Series MXVSB
View More Specs

Products 6
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-400-123
View Documents
Superior Glove™
MXVSB/S
Small
Pair for $47.75
Add to cart
 
17-400-124
View Documents
Superior Glove™
MXVSB/M
Medium
Pair for $47.75
Add to cart
 
17-400-125
View Documents
Superior Glove™
MXVSB/L
Large
Pair for $47.75
Add to cart
 
17-400-126
View Documents
Superior Glove™
MXVSB/XL
X-Large
Pair for $47.75
Add to cart
 
17-400-127
View Documents
Superior Glove™
MXVSB/XXL
2X-Large
Pair for $17.23
Add to cart
 
17-400-128
View Documents
Superior Glove™
MXVSB/3X
3X-Large
Pair for $47.75
Add to cart
 
Description

Description

  • Impact-resistant back-of-hand protection
  • Protects against knocks and bumps without sacrificing flexibility or dexterity
  • PVC Sure Grip™ palm patches enhance grip and improve fingertip protection
  • Reinforced thumb patch prevents degradation in high-stress areas
  • Adjustable cuffs for additional protection and comfort
Specifications

Specifications

Yellow, Black
ANSI A2
Mechanics, Construction, Oil/Gas Drilling and Material Handling
MXVSB
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit