Learn More
Superior Glove™ Contender™ Terry-Knit Metal-Stamping Gloves
Terry-knit aramid fiber gloves provide abrasion and cut protection. Ideal especially for metal stamping applications, but can also be used in automotive, assembly and material handling industries.
$28.30
Specifications
Certifications/Compliance ANSI 5 Heat
ANSI Abrasion Rating ANSI 2
Cut Resistance Rating ANSI A4
Material Aramid Fiber, Stainless Steel, Nylon
Coating Material Uncoated
View More Specs
Products 3
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-401-262
View Documents
Superior Glove™
TRFGK/M
Medium
Pair for $28.30
Pack of 6 PR for $161.50
Add to cart
17-401-263
View Documents
Superior Glove™
TRFGK/L
Large
Pair for $28.30
Pack of 6 PR for $161.50
Add to cart
17-401-264
View Documents
Superior Glove™
TRFGK/XL
X-Large
Pair for $28.30
Pack of 6 PR for $161.50
Add to cart
Description
Description
  • Terry knit contains composite filament fiber for abrasion and cut protection
  • Terry loops enhance cut resistance
  • Longer knit-wrist cuffs are fitting for stay-in-place comfort
  • Cotton blend absorbs surface oil/grease to increase grip on metal surfaces
Specifications
Specifications
ANSI 5 Heat
ANSI A4
Uncoated
ANSI 2
Aramid Fiber, Stainless Steel, Nylon
White