Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ CutBan™ Stockinette Cut-Resistant Sleeves

$6.63 - $9.63

Specifications

Cuff Style Knit
Color Black
Construction 2-ply, tubular knitting with high tenacity filament Yarn
Material Yarn
For Use With (Application) Metal handling, automotive, Metal fabrication
View More Specs

Products 3
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity & Availability  
17-400-152
View Documents
Superior Glove™
KPW22
22 in. 55.8 cm
Pair for $9.63
Dozen of 12 PR for $110.00
Add to cart
 
17-400-151
View Documents
Superior Glove™
KPW18
18 in. 45.7 cm
Pair for $8.73
Dozen of 12 PR for $99.80
Add to cart
 
17-400-150
View Documents
Superior Glove™
KPW12
12 in. 30.4 cm
Pair for $6.63
Dozen of 12 PR for $75.80
Add to cart
 
Description

Description

Specifications

Specifications

Knit
2-ply, tubular knitting with high tenacity filament Yarn
Metal handling, automotive, Metal fabrication
Black
Yarn
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit