Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Dexterity™ Nitrile Dipped Work Gloves
SDP

Catalog Number 17401286
Click to view available options
Size:
6
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Seamless knit cotton shell is comfortable and flexible, with a special additive in the nitrile coating to make gloves 100% biodegradable.

  • Nitrile dipped palm coating offers abrasion and puncture resistance
  • An additive in the coating makes gloves 100% biodegradable
  • Seamles knit cotton shell is comfortable and flexible
  • CFIA compliant
ANSI Abrasion Rating ANSI 4
Construction Seamless Knit
Coating Material Nitrile
Color Blue, White
Hand Specific
Material Cotton
Product Type Dip-coated Gloves
Size 6
For Use With (Application) Automotive Assembly, Maintenance, and Metal Stamping
Glove Cuff Style Knit
Puncture Rating ANSI/ISEA 105-16 Level 2
Series S15NT
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit