Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Superior Glove™ Dexterity™ Cut-Resistant String Knit Gloves, Nitrile Coating
Click to view available options
Size:
6
7
8
9
11
Description
- Combination of Kevlar™ and composite filament fiber provides 360° cut protection
- Nitrile palm coating offers secure dry grip and durability
- Ergonomically designed for unsurpassed comfort and dexterity
- Cut, abrasion and puncture resistant
Specifications
ANSI Abrasion Rating | ANSI 3 |
Coating Material | Nitrile Foam |
Color | Cream, Black |
Cut Resistance Rating | ANSI A3 |
Material | Dexterity |
Product Type | Cut-resistant Gloves |
Size | 6 |
For Use With (Application) | Automotive, light metal stamping, small parts handling, maintenance jobs, HVAC, glass |
Glove Cuff Style | Knit |
Model | S13KFNT/6 |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction