Superior™ Glove Emerald CX™ Composite Knit Gloves with Polyurethane Palms

Manufacturer: SUPERIOR GLOVE WORKS LTD S13CXPU/10

 View more versions of this product

Catalog No. 17-401-236

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Knit
    For Use With (Application) sharp Steel, circuit boards, automotive, Glass industry, Metal fabrication, assembly, sheet Metal work, HVAC
    Material Kevlar™, Stainless Steel
    Size 10
    Weight Light weight
    Coating Material Polyurethane
    Color Gray
    Cut Resistance Rating Level 4

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit