Learn More
Superior Glove™ Emerald CX™ Aramid Fiber Blend Gloves, Polyurethane Palm Coating
Cut-resistant gloves with a knit blend of aramid fiber and stainless steel. Polyurethane palm coating provides dexterity and tactility.
$7.79 - $21.10
Specifications
ANSI Abrasion Rating ANSI 3
Cut Resistance Rating ANSI A4
Puncture Rating ANSI/ISEA 105-16 Level 3
Material Aramid Fiber, Stainless Steel
Coating Material Polyurethane
View More Specs
Products 7
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-401-232
View Documents
Superior Glove™
S13CXPU/6
6
Pair for $21.10
Dozen of 12 PR for $241.00
Add to cart
17-401-233
View Documents
Superior Glove™
S13CXPU/7
7
Pair for $7.79
Dozen of 12 PR for $116.33
Add to cart
17-401-234
View Documents
Superior Glove™
S13CXPU/8
8
Pair for $7.79
Dozen of 12 PR for $116.33
Add to cart
17-401-235
View Documents
Superior Glove™
S13CXPU/9
9
Pair for $21.10
Dozen of 12 PR for $241.00
Add to cart
17-401-236
View Documents
Superior Glove™
S13CXPU/10
10
Pair for $21.10
Dozen of 12 PR for $241.00
Add to cart
17-401-237
View Documents
Superior Glove™
S13CXPU/11
11
Pair for $7.79
Dozen of 12 PR for $116.33
Add to cart
17-401-238
View Documents
Superior Glove™
S13CXPU/12
12
Pair for $21.10
Dozen of 12 PR for $241.00
Add to cart
Description
Description
  • Knitted with a blend of aramid fiber and stainless steel for high cut resistance
  • 13-gauge knit provides a balance between high dexterity and longevity
  • Polyurethane palm coating offers dexterity and tactile sensitivity while providing comfort and secure grip
  • Ideal for glass handling, non-marring gloves will not leave residue, smudges or fingerprints
  • Speckled color hides dirt for longer life span
Specifications
Specifications
ANSI 3
ANSI/ISEA 105-16 Level 3
Polyurethane
13
ANSI A4
Aramid Fiber, Stainless Steel
Gray