Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Superior Glove™ Emerald CX™ Cut-Resistant Gloves, Leather Palm
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Description
- Aramid fiber/stainless steel composite yarn blended with polyester
- Comfortable and durable
- 7-gauge, leather palm
- Ideal for moderate cut applications such as handling metal stampings, glass or other sharp materials
Specifications
Certifications/Compliance | ANSI 3 Heat |
Coating Material | Leather |
Color | Yellow, Black, Gray |
Cut Resistance Rating | ANSI A3 |
Material | Aramid Fiber, Stainless Steel Fiber |
Product Type | Cut-resistant Gloves |
Size | X-Small |
For Use With (Application) | Sheet metal, metal stamping and fabrication, glass handling |
Glove Cuff Style | Knit |
Model | SKWCPLP/XS |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction