Superior™ Glove Emerald CX Lite™ Cut-Resistant Gloves

Manufacturer: SUPERIOR GLOVE WORKS LTD S13KBFNT/8

 View more versions of this product

Catalog No. 17-401-217

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Knit
    For Use With (Application) HVAC, automotive, sharp Steel, Glass, Metal fabrication, light Metal stamping
    Material Nylon, Stainless Steel
    Size 8
    Coating Material Foamed Nitrile Coated Palms
    Color Black
    Cut Resistance Rating Level 2

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit