Superior™ Glove Sterile Cleanroom Cut-Resistant Knit Gloves

Cleanroom processed to minimize contamination from dust and particles for industry uses in pharmaceutical, electronic assembly and automotive paint line

Manufacturer: SUPERIOR GLOVE WORKS LTD CRSSXPUQ10

 View more versions of this product

Catalog No. 17-489-629

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Glove Cuff Style Knit
    For Use With (Application) cleanroom environment
    Size 10
    Sterile Yes
    Color White

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit