Learn More
Superior Glove™ Superior Touch™ Dyneema™ Gloves, Foam Nitrile Palm
Speckled gray Dyneema™ cut-resistant gloves feature a foam nitrile palm for secure grip when working with liquids and oils.
$8.73 - $25.25
Specifications
ANSI Abrasion Rating ANSI 4
Cut Resistance Rating ANSI A2
Puncture Rating ANSI/ISEA 105-16 Level 3
Material Dyneema™
Coating Material Nitrile foam
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-400-179
View Documents
Superior Glove™
S13SXGBFN/6
6
Pair for $8.73
Dozen of 12 PR for $226.66
Add to cart
17-400-185
View Documents
Superior Glove™
S13SXGBFN/12
12
Pair for $25.25
Dozen of 12 PR for $288.50
Add to cart
Description
Description
  • Speckled gray, 13-gauge gloves
  • Made of knit with lint-free, continuous-filament Dyneema™
  • Foam nitrile improves grip by absorbing and displacing liquids and oils on smooth objects
  • Gloves are washable and can be bleached at cold temperatures only
Specifications
Specifications
ANSI 4
ANSI/ISEA 105-16 Level 3
Nitrile foam
13
ANSI A2
Dyneema™
Gray, Black