Learn More
Superior Glove™ Sure-Grip™ Hot Mill Gloves
Specifications
Certifications/Compliance ASTM
For Use With (Application) Glass foundries, Plastics, material handling, automotive, castings, Steel mills, fabrication
Liner Material Acrylic
Temperature (English) Max. 392°F
Temperature (Metric) Max 200°C
View More Specs
Products 1
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-401-297
View Documents Promotion Details
Superior Glove™
SKC/ANTS/L
Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Specifications
Specifications
ASTM
Acrylic
200°C
Glass foundries, Plastics, material handling, automotive, castings, Steel mills, fabrication
392°F
Aramid Fiber, Cotton, Nitrile