Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Superior Glove™ TenActiv™ Composite Knit Cut-Resistant Gloves, Polyurethane Coating
Click to view available options
Size:
5
6
7
8
9
10
11
12
Description
- Continuous filament yarn is stronger than steel on an equal-weight basis
- 1250 grams of cut protection
- Lint-free
Withstands oils and solvents
Specifications
Coating Material | Polyurethane |
Color | Gray |
Cut Resistance Rating | ANSI A2 |
Material | Composite Fiber |
Product Type | Cut-resistant Gloves |
Size | 5 |
For Use With (Application) | Handling sharp objects, Glass plants, construction, automotive industry, aerospace industry, Metal fabrication, Metal stamping |
Glove Cuff Style | Knit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction