Superior Uniform™ Tugs Carnegie Slip-Resistant Boots
Manufacturer: Superior Uniform Group Inc 9038M10/L11.5
 View more versions of this product
Catalog No. 19-148-919
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Certifications/Compliance ASTM F2413 05M 1/75 C/75 EH
    Footbed Material Polyurethane
    Gender Unisex
    Waterproof Yes
    Color Black
    Material Leather
    Size 10 (Men's), 11.5 (women's)
    Sole Features Slip resistant Rubber outsole, Polyurethane midsole, oil, Water resistant sole
    Toe Material Steel
  • Description & Specifications
Related Products
Product Content Feedback