Superior Uniform™ Tugs Carnegie Slip-Resistant Boots
Manufacturer: Superior Uniform Group Inc 9038M14
 View more versions of this product
Catalog No. 19-148-924
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance ASTM F2413 05M 1/75 C/75 EH
    Footbed Material Polyurethane
    Gender Male
    Sole Features Slip resistant Rubber outsole, Polyurethane midsole, oil, Water resistant sole
    Toe Material Steel
    Waterproof Yes
    Color Black
    Material Leather
    Size 14
  • Description & Specifications
Related Products
Product Content Feedback