Learn More
Cell Biolabs IncSupplier Diversity Partner SUPEROXIDE DISMUTSE ASSY TR SZ  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner STA340T
The OxiSelect Superoxide Dismutase Activity Assay (Trial Size) provides fast, reliable measurement of SOD activity from plasma, serum, and cell or tissue lysates. 20 assays/kit including standards and unknowns.
Catalog No. 50-673-532