Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Superpack Pipette Tips

Gives superior results, optimal yield, and minimal sample loss compared to inferior pipette tips. Superpack Pipette Tips are available in bulk or in 10 x 96 sterile or non-sterile racks.

$63.75 - $134.10

Specifications

Color Natural
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Sterility Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Sterility Price Quantity & Availability  
MP0977987C2
View Documents
MP Biomedicals™
0977987C2
1000/Box NonSterile
Each for $63.75
Add to cart
 
MP0977987H2
View Documents
MP Biomedicals™
0977987H2
10 x 96/Box NonSterile
Each for $129.90
Add to cart
 
MP0977988H2
View Documents
MP Biomedicals™
0977988H2
10 x 96/Box Sterile
Each for $134.10
Add to cart
 
Description

Description

Superior pipette tips

  • Autoclavable
  • Recyclable
  • Very low coefficient of friction
  • Naturally hydrophobic
  • Metal-free
Specifications

Specifications

Natural
SDS
Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit