Learn More
Invitrogen™ SuperScript™ IV One-Step RT-PCR System with ezDNase™ Non-distribution product as customer accommodation.
Enabling faster, better RT-PCR.
$340.00 - $952.00
Includes:
  • SuperScript IV RT Mix, 12.5µL
  • 2X Platinum SuperFi RT-PCR Master Mix, 625µL
  • ezDNase Enzyme, 25µL
  • 10X ezDNase Buffer, 100µL
  • Nuclease-free water, 1.25mL
Products 2
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
12-595-025 Non-distribution product as customer accommodation.
View Documents
Invitrogen™
12595025
25
Each for $340.00
Add to cart
12-595-100 Non-distribution product as customer accommodation.
View Documents
Invitrogen™
12595100
100
Each for $952.00
Add to cart
Description
Description
The Invitrogen™ SuperScript™ IV One-Step RT-PCR System with ezDNase combines the high processivity Invitrogen™ SuperScript™ IV Reverse Transcriptase (RT) and the high-fidelity Invitrogen™ Platinum™ SuperFi™ DNA Polymerase to provide unmatched one-step RT-PCR performance. Using this convenient one-step formulation, you can perform both cDNA synthesis from total RNA or mRNA and PCR amplification in a single tube, using gene-specific primers.
  • Two-phase, hot-start activation mechanism for high specificity, yield, and room-temperature setup
  • Superior performance with sensitivity down to 0.01 pg of RNA, target length up to 13.8 kb, and the fastest one-step RT-PCR protocol
  • Reliable target detection even with RNA samples of suboptimal purity
  • Fast and easy gDNA removal for superior accuracy and confidence in your results
SDS
For research use only. Not for diagnostic procedures.