Learn More
MilliporeSigma™ Supershpher™ 100 RP-18 (4µm) Endcapped LiChroCART™ HPLC Cartridge
Yields an excellent pressure/separation performance ratio. MilliporeSigma Supershpher™ 100 RP-18 (4µm) Endcapped LiChroCART™ HPLC Cartridge is a reliable, versatile spherical silica gel carrier with reversed phase properties.
$633.56 - $716.68
Specifications
Column Type LiChroCART™ HPLC cartridge
Stationary Phase RP18
Particle Size 4 µm
View More Specs
Products 2
Catalog Number Mfr. No. Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Price Quantity & Availability  
M1519090001 View Documents MilliporeSigma™
1.51909.0001
125 mm Each for $633.56
Add to cart
M1519100001 View Documents MilliporeSigma™
1.51910.0001
250 mm Each for $716.68
Add to cart
Specifications
Specifications
LiChroCART™ HPLC cartridge
RP18
4 µm