Learn More
Fisherbrand™ Superslip Cover Slips
Make your automatic cover slip machines run smoothly
 View more versions of this product
Catalog No. 12-545-89P
Add to cart
Description
Description
  • Built-in moisture resistance
  • Excellent for use in humid environments
Specifications
Specifications
Glass
60 mm
24 x 60 mm
Superslip Cover Slip No. 1
Rectangular
24 mm
0.13 to 0.17 mm
1 oz./Pk.