Learn More
SUPT16H Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 205511AP
Catalog No. 50-173-2956
Add to cart
Description
Description
The antibody is specific to SUPT16H.
Transcription of protein-coding genes can be reconstituted on naked DNA with only the general transcription factors and RNA polymerase II. However, this minimal system cannot transcribe DNA packaged into chromatin, indicating that accessory factors may facilitate access to DNA. One such factor, FACT , interacts specifically with histones H2A/H2B to effect nucleosome disassembly and transcription elongation. FACT is composed of an 80 kDa subunit and a 140 kDa subunit; this gene encodes the 140 kDa subunit.
Specifications
Specifications
SUPT16H
0.43 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q920B9, Q9Y5B9
CDC68, FACT 140 kDa subunit, FACT complex subunit SPT16, FACT140, FACTP140, hSPT16, SPT16/CDC68, SUPT16H
Rabbit
IgG
150 μL
-20°C
Primary
11198, 114741
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
SUPT16H
Liquid
SUPT16H
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse