Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUPT3H Mouse anti-Human, Clone: OTI3D1, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Supplier: OriGene CF808117
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The transcription of many RNA polymerase II-dependent genes requires Spt3, a member of the S. cerevisiae SAGA complex. Transcription from delta sequences, the long terminal repeats that flank yeast Ty elements, requires the yeast SPT3 gene. Spt3 and Spt20 work together to recruit TATA-box binding protein (TBP) to the core promoter allowing TBP to bind to SAGA-dependent promoters. Null mutations in the Spt3 gene cause defects in sporulation, diploid filamentous growth, and haploid invasive growth, indicating that Spt3 has an important role in both mating and development pathways in yeast. At the promoters of some genes including yeast HO, HIS3 and TRP3 genes, Spt3 inhibits binding of TBP, resulting in reduced transcription. This repressive effect of Spt3 can be overcome by another member of the SAGA complex, GCN5, which promotes the formation of a TBP/TFIIA complex by histone acetylation.Specifications
SUPT3H | |
Monoclonal | |
Unconjugated | |
SUPT3H | |
SPT3, SPT3L | |
Mouse | |
Affinity Chromatography | |
RUO | |
8464 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3D1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O75486 | |
SUPT3H | |
Full length human recombit protein of human SUPT3H produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction