Learn More
SUPT4H1 Mouse anti-Human, Clone: OTI6B10, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF803430
Catalog No. 50-166-6488
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Component of the DRB sensitivity-inducing factor complex (DSIF complex), which regulates mRNA processing and transcription elongation by RNA polymerase II. DSIF positively regulates mRNA capping by stimulating the mRNA guanylyltransferase activity of RNGTT/CAP1A. DSIF also acts cooperatively with the negative elongation factor complex (NELF complex) to enhance transcriptional pausing at sites proximal to the promoter. Transcriptional pausing may facilitate the assembly of an elongation competent RNA polymerase II complex. DSIF and NELF promote pausing by inhibition of the transcription elongation factor TFIIS/S-II. TFIIS/S-II binds to RNA polymerase II at transcription pause sites and stimulates the weak intrinsic nuclease activity of the enzyme. Cleavage of blocked transcripts by RNA polymerase II promotes the resumption of transcription from the new 3 ft. terminus and may allow repeated attempts at transcription through natural pause sites. DSIF can also positively regulate transcriptional elongation and is required for the efficient activation of transcriptional elongation by the HIV-1 nuclear transcriptional activator, Tat. DSIF acts to suppress transcriptional pausing in transcripts derived from the HIV-1 LTR and blocks premature release of HIV-1 transcripts at terminator sequences.
Specifications
Specifications
SUPT4H1
Monoclonal
Unconjugated
SUPT4H1
Lyophilized
SUPT4H1
Full length human recombit protein of human SUPT4H1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI6B10
PBS with 8% trehalose and no preservative; pH 7.3
P63272
SPT4, SPT4H, SUPT4H, Supt4a
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
6827