Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Supt4h1 Polyclonal Antibody
Supt4h1 Polyclonal Antibody
Supplier: Thermo Scientific PA575694
Description
Supt4h1 Polyclonal Antibody for Western Blot

Specifications
Supt4h1 | |
Polyclonal | |
Unconjugated | |
SUPT4H1 | |
AL022777; DRB sensitivity-inducing factor 14 kDa subunit; DRB sensitivity-inducing factor small subunit; DRB sensitivity-inducing factor small subunit 1; DSIF p14; DSIF small subunit; DSIF small subunit 1; hSPT4; small hSpt4 subunit; SPT4; SPT4 homolog, DSIF elongation factor subunit; SPT4H; suppressor of Ty 4 homolog 1; suppressor of Ty 4 homolog 1 (S. cerevisiae); suppressor of Ty 4 homolog 2; suppressor of Ty 4A; Supt4a; Supt4h; Supt4h1; Supt4h1a; Supt4h2; transcription elongation factor SPT4; Transcription elongation factor SPT4 1; transcription elongation factor SPT4-A; zgc:92824; zgc:92824 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20922, 287608, 6827 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P63271, P63272 | |
Supt4a, SUPT4H1 | |
Synthetic peptide corresponding to residues in Human SUPT4H1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction