Learn More
SUR1 Mouse anti-Human, Mouse, Rat, Hamster, DyLight 755, Clone: S289-16, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259320IR
Catalog No. NBP259320IR
Add to cart
Description
Description
SUR1 Monoclonal antibody specifically detects SUR1 in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
SUR1
S289-16
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
ABCC8
Fusion protein amino acids 1548-1582 (cytoplasmic C-terminus) of rat SUR1
Protein G purified
ABC Transporters, Lipid and Metabolism, Nutrient Sensing in the Brain
Primary
6833
Human, Mouse, Rat, Hamster
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
ABC36, ATP-binding cassette, sub-family C (CFTR/MRP), member 8, HHF1Sulfonylurea receptor 1, HI, PHHIHRINS, sulfonylurea receptor (hyperinsulinemia), SUR1MRP8, SURATP-binding cassette transporter sub-family C member 8, TNDM2ATP-binding cassette sub-family C member 8
Mouse
IgG1
100 ul
Store at 4C in the dark.
Monoclonal
Detects 160kDa. Does not cross-react with SUR2B.