Learn More
Fisherbrand™ SURE-VUE™ Automated RPR Test Kits and Accessories
SURE-VUE™ AUTOMATED RPR (Rapid Plasma Reagin) Test for syphilis is a qualitative nontreponemal flocculation test for the detection of reagin antibodies in human serum and plasma as a screening test for serological evidence of syphilis for use on the ASI Evolution™ Analyzer.
 View more versions of this product
Catalog No. 23-038-081
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
For professional use only.
Specifications
Specifications
480 Tests/Pk.
Test Kit