Learn More
Fisherbrand™ SURE-VUE™ Automated RPR Test Kits and Accessories
SURE-VUE™ AUTOMATED RPR (Rapid Plasma Reagin) Test for syphilis is a qualitative nontreponemal flocculation test for the detection of reagin antibodies in human serum and plasma as a screening test for serological evidence of syphilis for use on the ASI Evolution™ Analyzer.
 View more versions of this product
Catalog No. 23-038-085
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. View substitute products »
This item is not returnable. View return policy
Description
Description
For professional use only.
Specifications
Specifications
Syphilis testing
3 x 5 mL/Pk.
Control Set