Learn More
Fisher Healthcare™ Sure-Vue™ Rubella Test Kit

A rapid latex particle agglutination test for the qualitative and semiquantitative determination of rubella virus antibodies in serum.
Specifications
CE Marker | Self-declared |
---|---|
Clia Complexity | Moderate |
Detectable Analytes | Rubella Virus Antibodies |
Detection Method | Latex Agglutination |
Sample Type | Serum |
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
23-038-183
![]() |
30 Pk. |
Pack of 30 for $568.00
|
N/A | ||||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-038-184
![]() |
100 Pk. |
Pack of 100 for $883.00
|
N/A | ||||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-038-185
![]() |
500 Pk. |
Pack of 500 for $3,554.00
|
N/A | ||||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
The Fisher Healthcare™ Sure-Vue™ Rubella Test is a rapid latex particle agglutination test for the qualitative and semiquantitative determination of rubella virus antibodies in serum.
- Excellent reproducibility and outstanding accuracy
- 3 levels of liquid with ready-to-use controls included
- White latex agglutination test
- Correlation with gold standard Hemagglutination Inhibition Test (HAI)
- Long shelf life
- Made in the USA
Specifications
Self-declared | |
Rubella Virus Antibodies | |
Serum | |
Rubella Test | |
Reagent, Dilution Buffer, Controls, Test Cards, Disposable Stirrer Pipets | |
Sure-Vue™ | |
Non-sterile |
Moderate | |
Latex Agglutination | |
1 to 2 IU/mL (Undiluted) or 10 to 20 IU/mL (Diluted) | |
Inert Latex Particles Coated with Gamma Globulins in 0.1% Sodium Azide | |
9 Wells | |
>99% | |
8 min |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.