Learn More
Fisherbrand™ SureFit™ Ergo-F Pipet Tips
Designed specifically for use with Fisherbrand pipetters
$182.90
Specifications
Filtered Filtered
View More Specs
Products 1
Catalog Number Description Volume (Metric) Tip Style Price Quantity & Availability  
Catalog Number Description Volume (Metric) Tip Style Price Quantity & Availability  
02-681-475
View Documents
100 to 1000 μL
Pack of 1000 for $182.90
Case of 5 PK for $823.50
Add to cart
Description
Description
Designed specifically for use with Fisherbrand pipetters
  • Available in three capacities, with or without aerosol barrier
  • Unique wall design provides an extra-strong seal specific to Fisherbrand pipetters
  • Require minimal ejection force
  • Certified by independent laboratory – Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
Specifications
Specifications
Filtered