Learn More
Seracare Life Sciences IncSupplier Diversity Partner SURELINK AP CONJUGATION KIT  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 56100026
SureLINK AP Conjugation Kits enables antibody or protein labeling with alkaline phosphatase (AP) in only 3 hours, saving you several hours over standard maleimide/thiol methods. This kit contains all reagents required to conjugate your protein or antibody, including ready-to-use buffers and an optimized storage buffer. Modified AP is supplied ready-to-use and requires no activation or purification. Sufficient reagents are provided to conjugate 3 samples, each containing between 0.1 -0.5 mg of antibody or protein.
Catalog No. 50-675-96