Learn More
Seracare Life Sciences IncSupplier Diversity Partner SURELINK CHROMOPHORIC BIOTIN  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 56100027
SureLINK Chromophoric Biotin Labeling Kits provide a quick and reliable method for labeling antibodies and other proteins with biotin and enable easy measurement of biotin incorporation in the conjugate. Proteins and peptides can be readily labeled with biotin through the modification of primary amines. The advantages of SureLINK Chromophoric Biotin for quantitating biotin incorporation contrast sharply with the limitations of the standard HABA method.
Catalog No. 50-675-99