Learn More
Seracare Life Sciences IncSupplier Diversity Partner SURELINK CHROMOPHORIC BIOTIN  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 56200030
SureLINK Chromophoric Biotin provides a quick and reliable method for labeling antibodies and other proteins with biotin. It also enables easy measurement of biotin incorporation in the conjugate. Proteins and peptides can be readily labeled with biotin through the modification of primary amines. The advantages of SureLINK Biotin for quantitating biotin incorporation contrast sharply with the limitations of the standard HABA method. Quantity is sufficient for labeling approximately 10 mg antibody or other proteins. SureLINK Biotin is stable for one year when stored at 2deg. C.
Catalog No. 50-676-00