Learn More
Seracare Life Sciences IncSupplier Diversity Partner SURELINK FITC LABELING KIT  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 56100020
SureLINK Fluorescein (FITC) Labeling Kit provides a quick and reliable method for labeling antibodies and other proteins with fluorescein isothiocyanate (FITC). The protocol is optimized and easy to follow, and enables labeling and purifying in just two hours with minimal hands-on time. Reagents are packaged in reaction-size vials with all the essential components for conjugation and purification provided in the kit, including spin filters to remove free dye after labeling. SureLINK Kits contain all the essential components for conjugation and purification. Kits are stable for at least one year when stored under recommended conditions.
Catalog No. 50-675-88