Learn More
Seracare Life Sciences IncSupplier Diversity Partner SURELINK HRP CONJUGATION KIT  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 56100024
SureLINK HRP Conjugation Kits are based on well-established periodate chemistry, yielding protein conjugates that are consistent and reproducible. This kit contains all reagents required to conjugate your protein or antibody, including ready-to-use buffers and an optimized storage buffer. Activated HRP is supplied ready-to-use and requires no activation or purification. Sufficient reagents are provided to conjugate 2 samples, each containing between 0.05 - 0.1 mg of antibody or protein.
Catalog No. 50-675-92