Learn More
Seracare Life Sciences IncSupplier Diversity Partner SURELINK MODIFIED AP  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 56200029
SureLINK Modified AP is a modified form of alkaline phosphatase (AP) that is ideal for conjugating AP to monoclonal antibodies or other proteins! AP has been modified with the linker succinimidyl 4-hydrazinonicotinate (SANH). Based on unique conjugation chemistry, the Modified AP is part of a two-step coupling reaction.The conjugation reaction takes approximately 2 hours to complete. Protein primary amine groups are covalently modified with a second linker succinimidyl-P-formyl benzoate (SFB). The SFB-Modified protein reacts through aldehyde reactive sites to a hydrazine group on the SANH-AP (Modified AP). No reducing agents are required for conjugation. The hydrazone bond formed between SFB and SANH is highly specific and stable.
Catalog No. 50-675-98