Fisherbrand™ SureOne Beveled Pipet Tips, Universal Fit: 1-200µL - PROMO
Universal-fit, beveled pipette tips offer ease of use, economical alternatives
 View more versions of this product
Catalog No. 02-707-450PM
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Product Type Bulk
    Material Polypropylene
    Length (Metric) 5cm
    Sterility Nonsterile
    Graduations 10, 50, 100μL
    Tip Style Beveled Ends
    Certifications/Compliance RNase, DNase, DNA-free
    Volume (Metric) 1 to 200μL
    Color Clear
  • Description & Specifications
Related Products
Product Content Feedback