Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SurePRIME CORE KIT 10

Chemically modified Taq DNA Polymerase + dNTPs in one kit

Manufacturer:  MP Biomedicals™ 11EPHSK105

Catalog No. MP1EPHSK105


Add to cart

Description

Description

  • Chemically modified Taq DNA Polymerase + dNTPs in one kit
  • SurePRIME™ CORE Kits contain all the reagents (presented as separate items) required for Hot Start PCR : MP Biomedicals SurePRIME™ DNA
  • Polymerase, high purity dNTPs Mix (>99%) and optimized incubation buffers.
  • Storage: -20°C
  • Units of SurePRIME™ DNA polymerase
Specifications

Specifications

chemically modified Taq DNA Polymerase + dNTPs in one kit
5 x 250
2X
Kits & Assays
Safety and Handling

Safety and Handling

SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.